Skip to Content
Merck
All Photos(1)

Documents

GF29539705

Hastealloy® c-22

rod, Ni59%/Cr22%/Mo13%/Fe 3%/W 3, 50.8 mm diameter, length 50 mm, temper annealed

Synonym(s):

NE097951

Sign Into View Organizational & Contract Pricing


About This Item

UNSPSC Code:
11101711
NACRES:
NA.23

form

rod

manufacturer/tradename

Goodfellow 295-397-05

L

50 mm

diameter

50.8 mm

Looking for similar products? Visit Product Comparison Guide

Related Categories

General description

For updated SDS information please visit www.goodfellow.com

Legal Information

Product of Goodfellow
Hastealloy is a registered trademark of HAYNES INTERNATIONAL, INC.

Certificates of Analysis (COA)

Search for Certificates of Analysis (COA) by entering the products Lot/Batch Number. Lot and Batch Numbers can be found on a product’s label following the words ‘Lot’ or ‘Batch’.

Already Own This Product?

Find documentation for the products that you have recently purchased in the Document Library.

Visit the Document Library

Our team of scientists has experience in all areas of research including Life Science, Material Science, Chemical Synthesis, Chromatography, Analytical and many others.

Contact Technical Service