GF29481396
Nickel-iron-molybdenum alloy, Ni80Fe15.5Mo4.5
200mm tube, 15mm outside diameter, 0.4mm wall thickness, 14.2mm inside diameter, as drawn
Synonym(s):
NJ277600
Sign Into View Organizational & Contract Pricing
All Photos(1)
About This Item
UNSPSC Code:
11101712
NACRES:
NA.23
form
tubes
manufacturer/tradename
Goodfellow 294-813-96
L × wall thickness
200 mm × 0.4 mm
O.D. × I.D.
15 mm × 14.2 mm
Looking for similar products? Visit Product Comparison Guide
General description
For updated SDS information please visit www.goodfellow.com.
Product of Goodfellow
Choose from one of the most recent versions:
Certificates of Analysis (COA)
Lot/Batch Number
It looks like we've run into a problem, but you can still download Certificates of Analysis from our Documents section.
If you need assistance, please contact Customer Support.
Already Own This Product?
Find documentation for the products that you have recently purchased in the Document Library.
Our team of scientists has experience in all areas of research including Life Science, Material Science, Chemical Synthesis, Chromatography, Analytical and many others.
Contact Technical Service