GF04734749
Vanadium
rod, 5.0 mm diameter, length 100 mm, purity 99.8%
Synonym(s):
V 007930
About This Item
Recommended Products
vapor pressure
8 mmHg ( 20 °C)
Assay
99.8%
form
rod
manufacturer/tradename
Goodfellow 047-347-49
resistivity
24.8-26.0 μΩ-cm, 20°C
L
100 mm
diameter
5.0 mm
bp
3380 °C (lit.)
mp
1890 °C (lit.)
density
6.11 g/mL at 25 °C (lit.)
SMILES string
[V]
InChI
1S/V
InChI key
LEONUFNNVUYDNQ-UHFFFAOYSA-N
Looking for similar products? Visit Product Comparison Guide
Related Categories
General description
Legal Information
Storage Class Code
13 - Non Combustible Solids
WGK
WGK 3
Flash Point(F)
Not applicable
Flash Point(C)
Not applicable
Certificates of Analysis (COA)
Search for Certificates of Analysis (COA) by entering the products Lot/Batch Number. Lot and Batch Numbers can be found on a product’s label following the words ‘Lot’ or ‘Batch’.
Already Own This Product?
Find documentation for the products that you have recently purchased in the Document Library.
Our team of scientists has experience in all areas of research including Life Science, Material Science, Chemical Synthesis, Chromatography, Analytical and many others.
Contact Technical Service