Sign In to View Organizational & Contract Pricing.
Select a Size
Change View
| Size/SKU | Availability | Price |
|---|
About This Item
Linear Formula:
C2H5OP(S)Cl2
CAS Number:
Molecular Weight:
179.01
NACRES:
NA.22
PubChem Substance ID:
UNSPSC Code:
12352100
EC Number:
216-100-4
MDL number:
Assay:
95%
To inquire about this product E23909, please contact your local Merck office or dealer. Contact Technical Service
vapor pressure
10 mmHg ( 55 °C)
Quality Segment
assay
95%
refractive index
n20/D 1.504 (lit.)
bp
55-68 °C/10 mmHg (lit.)
density
1.353 g/mL at 25 °C (lit.)
SMILES string
CCOP(Cl)(Cl)=S
InChI
1S/C2H5Cl2OPS/c1-2-5-6(3,4)7/h2H2,1H3
InChI key
SMODMOZAUKBGGM-UHFFFAOYSA-N
signalword
Danger
hcodes
Hazard Classifications
Acute Tox. 3 Dermal - Acute Tox. 3 Inhalation - Acute Tox. 4 Oral - Skin Corr. 1B
Storage Class
6.1A - Combustible acute toxic Cat. 1 and 2 / very toxic hazardous materials
wgk
WGK 3
flash_point_f
235.4 °F - closed cup
flash_point_c
113 °C - closed cup
ppe
Faceshields, Gloves, Goggles, type ABEK (EN14387) respirator filter
Choose from one of the most recent versions:
Already Own This Product?
Find documentation for the products that you have recently purchased in the Document Library.

