Skip to Content
Merck
All Photos(1)

Key Documents

DWK63A54

Kimble® Disposable Pasteur Pipet

L 5.75 in., soda-lime glass

Synonym(s):

63A54

Sign Into View Organizational & Contract Pricing


About This Item

UNSPSC Code:
41121500

material

soda-lime glass

packaging

pkg of 1000 ea

manufacturer/tradename

Kimble® 63A54

L

5.75 in.

Looking for similar products? Visit Product Comparison Guide

Application

Bacteriology, immunology, hematology, and serology

Features and Benefits

Removes risk of cross-contamination of high value specimens and laboratory reagents.

Legal Information

KIMBLE is a registered trademark of DWK Life Sciences

Choose from one of the most recent versions:

Certificates of Analysis (COA)

Lot/Batch Number

Sorry, we don't have COAs for this product available online at this time.

If you need assistance, please contact Customer Support.

Already Own This Product?

Find documentation for the products that you have recently purchased in the Document Library.

Visit the Document Library

Our team of scientists has experience in all areas of research including Life Science, Material Science, Chemical Synthesis, Chromatography, Analytical and many others.

Contact Technical Service