Skip to Content
Merck
All Photos(1)

Key Documents

D116009

Sigma-Aldrich

2,3-Dihydroxynaphthalene

98%

Synonym(s):

2,3-Naphthalenediol

Sign Into View Organizational & Contract Pricing


About This Item

Linear Formula:
C10H6(OH)2
CAS Number:
Molecular Weight:
160.17
Beilstein:
742375
EC Number:
MDL number:
UNSPSC Code:
12352100

Assay

98%

mp

161-165 °C (lit.)

SMILES string

Oc1cc2ccccc2cc1O

InChI

1S/C10H8O2/c11-9-5-7-3-1-2-4-8(7)6-10(9)12/h1-6,11-12H

InChI key

JRNGUTKWMSBIBF-UHFFFAOYSA-N

Looking for similar products? Visit Product Comparison Guide

Application

Reagent for synthesis of aromatic polyamides, crown ethers, W(VI) complexes, and sulfonic acids.

replaced by

Product No.
Description
Pricing

Pictograms

CorrosionExclamation mark

Signal Word

Danger

Hazard Statements

Hazard Classifications

Acute Tox. 4 Oral - Eye Dam. 1

Storage Class Code

11 - Combustible Solids

WGK

WGK 3

Flash Point(F)

347.0 °F

Flash Point(C)

175 °C

Personal Protective Equipment

dust mask type N95 (US), Eyeshields, Gloves

Choose from one of the most recent versions:

Certificates of Analysis (COA)

Lot/Batch Number

Don't see the Right Version?

If you require a particular version, you can look up a specific certificate by the Lot or Batch number.

Already Own This Product?

Find documentation for the products that you have recently purchased in the Document Library.

Visit the Document Library

Makromol. Chem., 194, 1595-1595 (1993)
Journal of the American Chemical Society, 115, 7916-7916 (1993)
Tetrahedron Letters, 34, 1287-1287 (1993)
J. Chem. Soc. Perkin Trans. II, 721-721 (1993)
Xiao-ming Lu et al.
Guang pu xue yu guang pu fen xi = Guang pu, 22(1), 36-38 (2003-08-28)
The title organo-molybdate derivatives are synthesized and their IR, 1H NMR spectra have been determined and the relations between the structures and the 1H NMR and IR parameters have been studied. The results indicate that the red shift of the

Our team of scientists has experience in all areas of research including Life Science, Material Science, Chemical Synthesis, Chromatography, Analytical and many others.

Contact Technical Service