Key Documents
CLS7732CO
Pyrex® gum stability glass bombs
replacement cover for gum bomb, reusuable, non-sterile, 1/cs
Synonym(s):
lab equipment accessory
About This Item
UNSPSC Code:
41121500
NACRES:
NB.43
Product CLS7732CO is not currently sold in your country. Contact Technical Service
material
borosilicate glass (borosilicate glass code 7740)
sterility
non-sterile
feature
reusable
packaging
case of 1
pack of 1 ×
manufacturer/tradename
Corning 7732-CO
O.D. × H
46 mm × 12 mm
suitability
suitable for (the determination of gum stability of gasoline)
General description
Note: Replacement covers for gum bomb CLS7732. The cover prevents condensed vapors in bomb stem from contaminating the sample, but does not interfere with free flow of oxygen. Made in accordance with ASTM D-525.
These PYREX® bombs are for the determination of gum stability of gasoline. The cover prevents condensed vapors in bomb stem from contaminating the sample, but does not interfere with free flow of oxygen. Made in accordance with ASTM D-525.
Legal Information
PYREX is a registered trademark of Corning, Inc.
Choose from one of the most recent versions:
Certificates of Analysis (COA)