CLS396520
Pyrex® aseptic filling bell
hose outlet O.D. 7.0 mm
Synonym(s):
Corning® 3965
Sign Into View Organizational & Contract Pricing
All Photos(1)
About This Item
Recommended Products
description
for use with tubes
manufacturer/tradename
Corning 3965-20
O.D. × H
28 mm × 75 mm
hose outlet O.D.
7.0 mm
Looking for similar products? Visit Product Comparison Guide
Related Categories
General description
For aseptic filling of vessels. Rubber tubing is attached to the top of the tubulation while the receiving vessel is placed inside the bell.
Legal Information
Corning is a registered trademark of Corning, Inc.
PYREX is a registered trademark of Corning, Inc.
Certificates of Analysis (COA)
Search for Certificates of Analysis (COA) by entering the products Lot/Batch Number. Lot and Batch Numbers can be found on a product’s label following the words ‘Lot’ or ‘Batch’.
Already Own This Product?
Find documentation for the products that you have recently purchased in the Document Library.
Our team of scientists has experience in all areas of research including Life Science, Material Science, Chemical Synthesis, Chromatography, Analytical and many others.
Contact Technical Service