Skip to Content
Merck

Skip To

CDS012016

3,4-difluorophenylacetyl chloride

AldrichCPR

Sign In to View Organizational & Contract Pricing.

Select a Size

Change View
Size/SKUAvailabilityPrice
100 mg
Please contact Customer Service for Availability
2 220,00 CZK

About This Item

Empirical Formula (Hill Notation):
C8H5ClF2O
Molecular Weight:
190.57
UNSPSC Code:
12352200
PubChem Substance ID:
MDL number:
NACRES:
NA.21

2 220,00 CZK


Please contact Customer Service for Availability

Request a Custom Order
Technical Service
Need help? Our team of experienced scientists is here for you.
Let Us Assist


form

solid

functional group

acyl chloride, fluoro

SMILES string

Fc1ccc(CC(Cl)=O)cc1F

InChI

1S/C8H5ClF2O/c9-8(12)4-5-1-2-6(10)7(11)3-5/h1-3H,4H2

InChI key

NHXZDVXZDDNPFM-UHFFFAOYSA-N

Other Notes

Please note that Sigma-Aldrich provides this product to early discovery researchers as part of a collection of unique chemicals. Sigma-Aldrich does not collect analytical data for this product. Buyer assumes responsibility to confirm product identity and/or purity. All sales are final.

NOTWITHSTANDING ANY CONTRARY PROVISION CONTAINED IN SIGMA-ALDRICH′S STANDARD TERMS AND CONDITIONS OF SALE OR AN AGREEMENT BETWEEN SIGMA-ALDRICH AND BUYER, SIGMA-ALDRICH SELLS THIS PRODUCT "AS-IS" AND MAKES NO REPRESENTATION OR WARRANTY WHATSOEVER WITH RESPECT TO THIS PRODUCT, INCLUDING ANY (A) WARRANTY OF MERCHANTABILITY; (B) WARRANTY OF FITNESS FOR A PARTICULAR PURPOSE; OR (C) WARRANTY AGAINST INFRINGEMENT OF INTELLECTUAL PROPERTY RIGHTS OF A THIRD PARTY; WHETHER ARISING BY LAW, COURSE OF DEALING, COURSE OF PERFORMANCE, USAGE OF TRADE OR OTHERWISE.

Compare Similar Items

View Full Comparison

Show Differences

1 of 1

This Item
CDS025263290424
description

AldrichCPR

description

AldrichCPR

description

95%

form

solid

form

-

form

-

functional group

fluoro, acyl chloride

functional group

bromo

functional group

fluoro, carboxylic acid


Storage Class

11 - Combustible Solids

wgk

WGK 3

flash_point_f

Not applicable

flash_point_c

Not applicable



Choose from one of the most recent versions:

Certificates of Analysis (COA)

Lot/Batch Number

It looks like we've run into a problem, but you can still download Certificates of Analysis from our Documents section.

If you need assistance, please contact Customer Support

Already Own This Product?

Find documentation for the products that you have recently purchased in the Document Library.

Visit the Document Library



Questions

Reviews

No rating value

Active Filters