Key Documents
BR90624
BRAND® glass beaker with spout, tall form
volume 100 mL, with graduation
Synonym(s):
Beakers, Glass
Select a Size
10 EA
CZK 2,630.00
CZK 2,630.00
Please contact Customer Service for Availability
Select a Size
Change View
10 EA
CZK 2,630.00
About This Item
UNSPSC Code:
41121803
NACRES:
NB.43
CZK 2,630.00
Please contact Customer Service for Availability
material
borosilicate glass 3.3
feature
with graduation
manufacturer/tradename
BRAND 90624
volume
100 mL
General description
The BRAND® Glass Beaker with Spout, tall form, is constructed from high-quality Boro 3.3 glass, ensuring durability and reliability in laboratory environments. Compliant with DIN EN ISO 3819 standards, this beaker features clear graduation markings for accurate liquid measurement. The tall form design provides increased capacity, while the integrated spout facilitates easy pouring, minimizing spills and enhancing precision during liquid transfer.
Other Notes
Browse all glass beakers
Legal Information
BRAND is a registered trademark of BRAND GMBH + CO KG
Choose from one of the most recent versions:
Certificates of Analysis (COA)