Skip to Content
Merck
All Photos(1)

Documents

BR781317

BRAND® PCR tubes, strips of 8

attached transparent single caps, low profile, capacity 0.15 mL, white wells

Synonym(s):

BIO-CERT, PCR, PCR Tube Strips, PCR Tubes, Tube Strips, Tubes

Sign Into View Organizational & Contract Pricing


About This Item

UNSPSC Code:
41121800
NACRES:
NB.22

material

polypropylene tube
white wells

description

DNase-/RNase- DNA-free

sterility

non-sterile

feature

neck thread push fit
attached transparent single caps

packaging

pack of 120

manufacturer/tradename

BRAND 781317

technique(s)

PCR: suitable

capacity

0.15 mL

working volume

0.15 mL

suitability

suitable for (for qPCR)

binding type

non-treated surface

Looking for similar products? Visit Product Comparison Guide

General description

PCR 8- and 12-tube strips offer a flexible solution for your PCR or qPCR. Domed caps provide extra sealing pressure via the lid of the thermal cycler. Flat caps are required for qPCR. Attached cap strips can be easily opened and closed with one hand. Separate cap and film strips are suitable for automation, as they won′t obstruct robotics. No matter which cap you choose, all variants seal the vessels reliably, effectively protecting your samples from evaporation and contamination. The extra-thin walls allow for even heat transfer during the process of heating and cooling for short cycle time.

Application

PCR, qPCR (flat caps only)

Features and Benefits

  • High flexibility with a wide range of cap types and film strips
  • Reduced evaporation losses thanks to tightly closing cap strips
  • Rapid heat transfer through thin vessel walls
  • Numbered wells for clear identification
  • Free of: DNA, RNA, DNase, RNase, pyrogens, PCR inhibitors

Legal Information

BRAND is a registered trademark of BRAND GMBH + CO KG

Certificates of Analysis (COA)

Search for Certificates of Analysis (COA) by entering the products Lot/Batch Number. Lot and Batch Numbers can be found on a product’s label following the words ‘Lot’ or ‘Batch’.

Already Own This Product?

Find documentation for the products that you have recently purchased in the Document Library.

Visit the Document Library

Our team of scientists has experience in all areas of research including Life Science, Material Science, Chemical Synthesis, Chromatography, Analytical and many others.

Contact Technical Service