Skip to Content
Merck
All Photos(1)

Key Documents

BR402046

BRAND® volumetric flask for sugar analysis

size 200 mL

Synonym(s):

Flasks, SILBERBRAND, Volumetric flasks

Sign Into View Organizational & Contract Pricing

Select a Size

2 EA
CZK 2,170.00

CZK 2,170.00


Please contact Customer Service for Availability


Select a Size

Change View
2 EA
CZK 2,170.00

About This Item

UNSPSC Code:
41121804
NACRES:
NB.43

CZK 2,170.00


Please contact Customer Service for Availability

material

borosilicate glass

feature

class B For Kohlrausch sugar analysis. Calibrated to contain (TC, In).

packaging

pack of 2 ea

manufacturer/tradename

BRAND 402046

size

200 mL

Looking for similar products? Visit Product Comparison Guide

General description

Borosilicate glass 3.3. For Kohlrausch sugar analysis. Error limits according to Class B. Calibrated to contain (TC, In.).

Legal Information

BRAND is a registered trademark of BRAND GMBH + CO KG

Choose from one of the most recent versions:

Certificates of Analysis (COA)

Lot/Batch Number

It looks like we've run into a problem, but you can still download Certificates of Analysis from our Documents section.

If you need assistance, please contact Customer Support.

Already Own This Product?

Find documentation for the products that you have recently purchased in the Document Library.

Visit the Document Library

Our team of scientists has experience in all areas of research including Life Science, Material Science, Chemical Synthesis, Chromatography, Analytical and many others.

Contact Technical Service