BR23809
BRAND® compact automatic SILBERBRAND burette, without support/clamp
volume 10 mL, with LDPE bottle and PVC filling tube
Synonym(s):
Automatic Burettes, Burettes, Compact Automatic Burettes, SILBERBRAND
All Photos(2)
About This Item
Recommended Products
material
AR-Glass
manufacturer/tradename
BRAND 23809
volume measuring range
10 mL, accuracy: ±0.05 mL
volume
10 mL
Related Categories
General description
The Modular Automatic Burette Concept! Fast to dismantle and easy to clean. Easy to repair - all individual components replaceable! Boro 3.3, detachable PTFE stopcock. Error limits according to class B, DIN EN ISO 385. Calibrated to deliver (TD, Ex).
Legal Information
BRAND is a registered trademark of BRAND GMBH + CO KG