BR130422
BRAND® square bottle with tamper-proof screw cap, PE-HD
capacity 1000 mL, wide-neck
Synonym(s):
Bottles, Plastics, Square bottles
All Photos(1)
About This Item
material
high-density polyethylene bottle
polypropylene cap
square bottle
translucent bottle
wide-mouth bottle
wide-neck
Joint
Joints threaded neck (GL54)
feature
with cap
packaging
pack of 1 ea
manufacturer/tradename
BRAND 130422
L × H
85 mm × 210 mm
capacity
1000 mL
General description
Translucent. Screw cap with sealing cone, PP, blue, with cadmium-free pigments. Tamper-proof closure with press-off retaining ring. Space-saving square base.
Legal Information
BRAND is a registered trademark of BRAND GMBH + CO KG