Skip to Content
Merck
All Photos(1)

Documents

BMSR2024EUK

Benchmark RotoMini Plus Variable Speed Rotator with tube holders

AC/DC input 230 V, UK plug

Synonym(s):

tube rotator, mini tube rotator, lab roller, mini lab roller, lab revolver, rotoflex, rotoflex, argos rotator, mini rotator

Sign Into View Organizational & Contract Pricing


About This Item

UNSPSC Code:
41103811
NACRES:
NB.13

feature

positions< 96

manufacturer/tradename

Benchmark Scientific R2024-E-UK

parameter

5-70 rpm speed

W × L × H

41 cm × 13.7 cm × 19.9 cm

Looking for similar products? Visit Product Comparison Guide

General description

The RotoMini series of rotating mixers provides a wide range of mixing capabilities, suitable for an array of laboratory applications including resuspension, protein binding, nucleic acid purification, histochemistry, hybridization etc. The mixers include interchangeable tube holders for accommodating all common tube sizes from 0.5ml to 50ml. and the motion can be adjusted from vertical (end over end) mixing, to horizontal (rolling or hybridization style) mixing. RotoMini rotators are available in the "Plus" configuration. The Plus models provide the following additional features: Variable speed: 5 to 70rpm "Mix" function: Rotating in combination w/ oscillations (increased mixing) "Pause" function: Rotating with intermittent "stops" during rotation "Rock" function: Back and forth, gentle rocking motion (seesaw) The Plus models also offer an additional rocking platform (4.5 x 3 in.), as well as a 96 x 0.2ml tube/strip/plate holder for mixing PCR vessels in the "Mix" (oscillating) mode.

Application

Mixing of samples in tubes

Certificates of Analysis (COA)

Search for Certificates of Analysis (COA) by entering the products Lot/Batch Number. Lot and Batch Numbers can be found on a product’s label following the words ‘Lot’ or ‘Batch’.

Already Own This Product?

Find documentation for the products that you have recently purchased in the Document Library.

Visit the Document Library

Our team of scientists has experience in all areas of research including Life Science, Material Science, Chemical Synthesis, Chromatography, Analytical and many others.

Contact Technical Service