Skip to Content
Merck
All Photos(1)

Key Documents

BMSE1200CS1

Benchmark Extra casting set for myGel ”InstaView”

for use with 10.5x10cm and 10.5x6cm gels

Synonym(s):

gel casting kit

Sign Into View Organizational & Contract Pricing


About This Item

UNSPSC Code:
41105300
NACRES:
NB.13

manufacturer/tradename

Benchmark Scientific E1200-CS1

compatibility

for use with 10.5x10cm and 10.5x6cm gels

General description

Casting set for myGel electrophroresis systems. Includes stand, trays, E1101COMB1 doublesided combs

Application

myGel, casting, electrophoresis, agarose

Choose from one of the most recent versions:

Certificates of Analysis (COA)

Lot/Batch Number

Sorry, we don't have COAs for this product available online at this time.

If you need assistance, please contact Customer Support.

Already Own This Product?

Find documentation for the products that you have recently purchased in the Document Library.

Visit the Document Library

Our team of scientists has experience in all areas of research including Life Science, Material Science, Chemical Synthesis, Chromatography, Analytical and many others.

Contact Technical Service