Key Documents
BAH379430000
SP Bel-Art® FEP Coated Steel Forceps
straight (fully coated)
Select a Size
2 EACH
CZK 2,510.00
CZK 2,510.00
Please contact Customer Service for Availability
Select a Size
Change View
2 EACH
CZK 2,510.00
About This Item
UNSPSC Code:
41122800
NACRES:
NB.43
CZK 2,510.00
Please contact Customer Service for Availability
material
straight (fully coated)
packaging
pkg of 2 ea
manufacturer/tradename
Bel-Art H37943-0000
L
11.4 cm (4½ in.) , long
General description
These forceps are fully coated with brown PTFE FEP and comply with FDA regulations concerning direct food contact.
Features and Benefits
  • Wide tips are perfect for lifting cover glasses
  • Available in two styles, bent or straight tip
Legal Information
Bel-Art is a registered trademark of SP Scienceware, a division of SP
Choose from one of the most recent versions:
Certificates of Analysis (COA)