Key Documents
BAH189620000
SP Bel-Art® Poxygrid® Microliter Pipettor Rack
to hold, 10 pipettors
Synonym(s):
Pipette Rack
Select a Size
1 EACH
CZK 4,270.00
CZK 4,270.00
Please contact Customer Service for Availability
Select a Size
Change View
1 EACH
CZK 4,270.00
About This Item
UNSPSC Code:
41122800
NACRES:
NB.43
CZK 4,270.00
Please contact Customer Service for Availability
feature
to hold 10 pipettors
packaging
pack of 1 ea
manufacturer/tradename
Bel-Art H18962-0000
L × W × H
9 in. (229 mm) × 5 in. (127 mm) × 6.75 in. (171 mm)
General description
Store pipettors while protecting pipettor tips in these epoxy-coated aluminum racks that provide quick, easy access for more efficient work.
Features and Benefits
  • Holds up to 10 round shape pipettors such as Finnpipette®, Socorex, Eppendorf® and other popular brands; two end holes fit Pipette-Aid pipette fillers
  • Two stepped rows of 5 each
  • Second lower support row has smaller holes to accept tapered ends of the pipettors to keep them vertical
Legal Information
Bel-Art is a registered trademark of SP Scienceware, a division of SP
Eppendorf is a registered trademark of Eppendorf AG
Finnpipette is a registered trademark of Thermo Fisher Scientific or its subsidiaries
Poxygrid is a registered trademark of SP Scienceware, a division of SP
Choose from one of the most recent versions:
Certificates of Analysis (COA)