Key Documents
BAF651000000
SP Bel-Art® Proculture Cordless Homogenizer unit for Micro-Tube Homogenizer System
includes cordless motor, 2 AA batteries and pestle adapter
Select a Size
1 EACH
CZK 3,430.00
CZK 3,430.00
Please contact Customer Service for Availability
Select a Size
Change View
1 EACH
CZK 3,430.00
About This Item
UNSPSC Code:
41101504
NACRES:
NB.11
CZK 3,430.00
Please contact Customer Service for Availability
packaging
pack of 1 ea
manufacturer/tradename
Bel-Art F65100-0000
battery power
2 AA
General description
Hand-held, cordless homogenizer features a simple push in / pull out pestle exchange and is ideal for thorough and faster processing of one to many samples. Matched pestle tips and micro tubes can be used either with the homogenizer or with hand processing.
Features and Benefits
  • Homogenize samples directly in their storage container
  • Obtain thorough mixtures with the tight pestle-to-microtube tolerance
  • Protect your samples—tubes and pestles are made from virgin polypropylene
Legal Information
Bel-Art is a registered trademark of SP Scienceware, a division of SP
Choose from one of the most recent versions:
Certificates of Analysis (COA)