Skip to Content
Merck
All Photos(1)

Key Documents

B32203

Sigma-Aldrich

S-Benzylthioglycolic acid

97%

Synonym(s):

(Benzylthio)acetic acid, Benzylmercaptoacetic acid

Sign Into View Organizational & Contract Pricing


About This Item

Linear Formula:
C6H5CH2SCH2CO2H
CAS Number:
Molecular Weight:
182.24
EC Number:
MDL number:
UNSPSC Code:
12352100
PubChem Substance ID:

Quality Level

Assay

97%

form

solid

mp

59-63 °C (lit.)

SMILES string

OC(=O)CSCc1ccccc1

InChI

1S/C9H10O2S/c10-9(11)7-12-6-8-4-2-1-3-5-8/h1-5H,6-7H2,(H,10,11)

InChI key

AWLVTQRRKPBQEQ-UHFFFAOYSA-N

Storage Class Code

13 - Non Combustible Solids

WGK

WGK 3

Flash Point(F)

Not applicable

Flash Point(C)

Not applicable

Personal Protective Equipment

dust mask type N95 (US), Eyeshields, Gloves

Choose from one of the most recent versions:

Certificates of Analysis (COA)

Lot/Batch Number

Don't see the Right Version?

If you require a particular version, you can look up a specific certificate by the Lot or Batch number.

Already Own This Product?

Find documentation for the products that you have recently purchased in the Document Library.

Visit the Document Library

H D Babenzien et al.
Zeitschrift fur allgemeine Mikrobiologie, 19(8), 527-533 (1979-01-01)
Dibenzylsulfid (DBS) as a model of the organic sulfur compounds in crude oil was converted by a mixed culture (containing Pseudomonas aeruginosa) into several water soluble organic substances. Whereas these compounds are detectable with DC- and IR-spectroscopic techniques, benzylmercaptoacetic acid

Questions

Reviews

No rating value

Active Filters

Our team of scientists has experience in all areas of research including Life Science, Material Science, Chemical Synthesis, Chromatography, Analytical and many others.

Contact Technical Service