Skip to Content
Merck
All Photos(2)

Key Documents

ALD00454

Sigma-Aldrich

2-(3,3-Diethyl-1-triazenyl)-4-methylbenzenesulfonyl chloride

Synonym(s):

2-[(1E)-3,3-diethyl-1-triazen-1-yl]-4-methyl-benzenesulfonyl chloride

Sign Into View Organizational & Contract Pricing


About This Item

Empirical Formula (Hill Notation):
C11H16ClN3O2S
CAS Number:
Molecular Weight:
289.78
MDL number:
UNSPSC Code:
12352000
PubChem Substance ID:
NACRES:
NA.22

Assay

97%

Quality Level

form

powder

availability

available only in USA

mp

76 °C

SMILES string

CCN(CC)\N=N\c1cc(C)ccc1S(Cl)(=O)=O

InChI

1S/C11H16ClN3O2S/c1-4-15(5-2)14-13-10-8-9(3)6-7-11(10)18(12,16)17/h6-8H,4-5H2,1-3H3/b14-13+

InChI key

PDGZSZLMBJSDQC-BUHFOSPRSA-N

Looking for similar products? Visit Product Comparison Guide

Application

This triazine-containing toluenesulfonyl chloride "portable desaturase" (Tz°Cl) is a bench-stable that is facile to install on alcohol and amine functionalities to direct selective desaturation of hydrocarbons at remote positions, while leaving behind a synthetically useful tosyl group.

Pictograms

FlameSkull and crossbones

Signal Word

Danger

Hazard Statements

Hazard Classifications

Acute Tox. 3 Oral - Self-react. C

Storage Class Code

5.2 - Organic peroxides and self-reacting hazardous materials

WGK

WGK 3

Flash Point(F)

Not applicable

Flash Point(C)

Not applicable


Choose from one of the most recent versions:

Certificates of Analysis (COA)

Lot/Batch Number

Don't see the Right Version?

If you require a particular version, you can look up a specific certificate by the Lot or Batch number.

Already Own This Product?

Find documentation for the products that you have recently purchased in the Document Library.

Visit the Document Library

Related Content

Baran Group offers zinc-based reagents for versatile transformations in organic synthesis.

Our team of scientists has experience in all areas of research including Life Science, Material Science, Chemical Synthesis, Chromatography, Analytical and many others.

Contact Technical Service