Skip to Content
Merck

97285

Z-D-Tyr-OH

≥98.0% (TLC)

Synonym(s):

Z-D-tyrosine

Sign In to View Organizational & Contract Pricing.

Select a Size

Change View

About This Item

Empirical Formula (Hill Notation):
C17H17NO5
CAS Number:
Molecular Weight:
315.32
UNSPSC Code:
12352200
NACRES:
NA.22
PubChem Substance ID:
MDL number:
Beilstein/REAXYS Number:
6071009
Technical Service
Need help? Our team of experienced scientists is here for you.
Let Us Assist


Quality Segment

assay

≥98.0% (TLC)

form

powder

application(s)

peptide synthesis

SMILES string

OC(=O)[C@@H](Cc1ccc(O)cc1)NC(=O)OCc2ccccc2

InChI

1S/C17H17NO5/c19-14-8-6-12(7-9-14)10-15(16(20)21)18-17(22)23-11-13-4-2-1-3-5-13/h1-9,15,19H,10-11H2,(H,18,22)(H,20,21)/t15-/m1/s1

InChI key

MCRMUCXATQAAMN-OAHLLOKOSA-N



Storage Class

13 - Non Combustible Solids

wgk

WGK 3

flash_point_f

Not applicable

flash_point_c

Not applicable

ppe

Eyeshields, Gloves, type N95 (US)



Choose from one of the most recent versions:

Certificates of Analysis (COA)

Lot/Batch Number

It looks like we've run into a problem, but you can still download Certificates of Analysis from our Documents section.

If you need assistance, please contact Customer Support

Already Own This Product?

Find documentation for the products that you have recently purchased in the Document Library.

Visit the Document Library