Skip to Content
Merck

96080

(Z-Cys-OH)2

≥98.0% (T)

Synonym(s):

Di-Z-L-cystine

Sign In to View Organizational & Contract Pricing.

Select a Size

Change View

About This Item

Empirical Formula (Hill Notation):
C22H24N2O8S2
CAS Number:
Molecular Weight:
508.56
PubChem Substance ID:
UNSPSC Code:
12352200
EC Number:
230-181-3
MDL number:
Beilstein/REAXYS Number:
2550640
Technical Service
Need help? Our team of experienced scientists is here for you.
Let Us Assist


assay

≥98.0% (T)

optical activity

[α]/D −91±2°, c = 2% in acetic acid

mp

118-122 °C (dec.)

SMILES string

OC(=O)[C@H](CSSC[C@H](NC(=O)OCc1ccccc1)C(O)=O)NC(=O)OCc2ccccc2



Storage Class

13 - Non Combustible Solids

WGK

WGK 3

Flash Point (°F)

Not applicable

Flash Point (°C)

Not applicable

PPE (personal protective equipment)

Eyeshields, Gloves, type N95 (US)



Choose from one of the most recent versions:

Certificates of Analysis (COA)

Lot/Batch Number

Don't see the Right Version?

If you require a particular version, you can look up a specific certificate by the Lot or Batch number.

Already Own This Product?

Find documentation for the products that you have recently purchased in the Document Library.

Visit the Document Library