Skip to Content
Merck
All Photos(1)

Key Documents

932809

Sigma-Aldrich

Tweezer

Style 51S, ESD Rubber Coated

Synonym(s):

Grippers, Tweezers

Sign Into View Organizational & Contract Pricing

Select a Size

1 EACH
CZK 2,670.00

CZK 2,670.00


Please contact Customer Service for Availability

Request a Bulk Order

Select a Size

Change View
1 EACH
CZK 2,670.00

About This Item

UNSPSC Code:
41111761
NACRES:
NB.35

CZK 2,670.00


Please contact Customer Service for Availability

Request a Bulk Order

Looking for similar products? Visit Product Comparison Guide

General description

EMS techware

Application

ESD Rubber Coated Tweezers, Style 51S can be used for safe ergonomic cushion grips (resistivity 10E9 Ohm) for handling ESD sensitive components or small static items. Soft ESD safe cushion grips (resistively 10E8 ohm), these tweezers are ergonomically designed.

OAL: 115 mm. Material: Anti-Acid/Anti-MagSS. Tips: Bent for maximum visibility extra fine.

Choose from one of the most recent versions:

Certificates of Analysis (COA)

Lot/Batch Number

It looks like we've run into a problem, but you can still download Certificates of Analysis from our Documents section.

If you need assistance, please contact Customer Support.

Already Own This Product?

Find documentation for the products that you have recently purchased in the Document Library.

Visit the Document Library

Questions

Reviews

No rating value

Active Filters

Our team of scientists has experience in all areas of research including Life Science, Material Science, Chemical Synthesis, Chromatography, Analytical and many others.

Contact Technical Service