Skip to Content
Merck
All Photos(1)

Key Documents

92803

Sigma-Aldrich

2,4,6-Trimethyl-2,4,6-trivinylcyclotrisilazane

technical, ≥90%

Sign Into View Organizational & Contract Pricing


About This Item

Empirical Formula (Hill Notation):
C9H21N3Si3
CAS Number:
Molecular Weight:
255.54
Beilstein:
2055568
EC Number:
MDL number:
UNSPSC Code:
12352103
PubChem Substance ID:

grade

technical

Assay

≥90%

form

liquid

refractive index

n20/D 1.482

density

0.964 g/mL at 20 °C (lit.)

SMILES string

C[Si]1(N[Si](C)(N[Si](C)(N1)C=C)C=C)C=C

InChI

1S/C9H21N3Si3/c1-7-13(4)10-14(5,8-2)12-15(6,9-3)11-13/h7-12H,1-3H2,4-6H3

InChI key

RFSBGZWBVNPVNN-UHFFFAOYSA-N

Storage Class Code

10 - Combustible liquids

WGK

WGK 3

Personal Protective Equipment

dust mask type N95 (US), Eyeshields, Gloves

Choose from one of the most recent versions:

Certificates of Analysis (COA)

Lot/Batch Number

Don't see the Right Version?

If you require a particular version, you can look up a specific certificate by the Lot or Batch number.

Already Own This Product?

Find documentation for the products that you have recently purchased in the Document Library.

Visit the Document Library

Customers Also Viewed

Slide 1 of 1

1 of 1

Our team of scientists has experience in all areas of research including Life Science, Material Science, Chemical Synthesis, Chromatography, Analytical and many others.

Contact Technical Service