Skip to Content
Merck
All Photos(1)

Documents

92509

Sigma-Aldrich

2-Cyclohexen-1-one

produced by BASF, ≥98.0% (GC)

Sign Into View Organizational & Contract Pricing


About This Item

Linear Formula:
C6H8(=O)
CAS Number:
Molecular Weight:
96.13
Beilstein:
1280477
EC Number:
MDL number:
UNSPSC Code:
12352100
PubChem Substance ID:
NACRES:
NA.22

grade

produced by BASF

vapor pressure

760 mmHg ( 168 °C)

Assay

≥98.0% (GC)

form

liquid

impurities

≤0.5% water

refractive index

n20/D 1.488 (lit.)

bp

171-173 °C (lit.)

mp

−53 °C (lit.)

density

0.993 g/mL at 25 °C (lit.)

SMILES string

O=C1CCCC=C1

InChI

1S/C6H8O/c7-6-4-2-1-3-5-6/h2,4H,1,3,5H2

InChI key

FWFSEYBSWVRWGL-UHFFFAOYSA-N

Looking for similar products? Visit Product Comparison Guide

Pictograms

FlameSkull and crossbones

Signal Word

Danger

Hazard Statements

Hazard Classifications

Acute Tox. 1 Inhalation - Acute Tox. 2 Dermal - Acute Tox. 3 Oral - Flam. Liq. 3

Storage Class Code

3 - Flammable liquids

WGK

WGK 3

Flash Point(F)

132.8 °F - closed cup

Flash Point(C)

56 °C - closed cup

Personal Protective Equipment

dust mask type N95 (US), Eyeshields, Gloves

Certificates of Analysis (COA)

Search for Certificates of Analysis (COA) by entering the products Lot/Batch Number. Lot and Batch Numbers can be found on a product’s label following the words ‘Lot’ or ‘Batch’.

Already Own This Product?

Find documentation for the products that you have recently purchased in the Document Library.

Visit the Document Library

Ronaldo Mariz et al.
Chemistry (Weinheim an der Bergstrasse, Germany), 16(48), 14335-14347 (2010-11-16)
A family of chiral C(2)-symmetric disulfoxide ligands possessing biaryl atropisomeric backbones has been synthesized by using the Andersen methodology. Complete characterization includes X-ray crystallographic studies of all ligands and some of their rhodium complexes. Their synthesis, optical purity, electronic properties
Xingwang Wang et al.
Journal of the American Chemical Society, 130(19), 6070-6071 (2008-04-22)
A highly enantioselective epoxidation of cyclic enones with hydrogen peroxide has been developed that is catalyzed by chiral primary amine salts.
Olivia Dean et al.
Pharmacology, biochemistry, and behavior, 97(2), 293-300 (2010-09-08)
Oxidative stress has been implicated in several psychiatric illnesses, including schizophrenia. Glutathione is the brain's primary antioxidant and decreased levels of brain glutathione are reported in schizophrenia. Prepulse inhibition (PPI) is a measure of sensory gating, and PPI is reduced
Marcin Kwit et al.
Organic & biomolecular chemistry, 8(24), 5635-5645 (2010-10-14)
The absolute configurations of 2-cyclohexenone cis-diol metabolites resulting from the biotransformation of the corresponding phenols have been determined by comparison of their experimental and calculated circular dichroism spectra (TDDFT at the PCM/B2LYP/Aug-cc-pVTZ level), optical rotations (calculated at the PCM/B3LYP/Aug-cc-pVTZ level)
Polyssena Renzi et al.
Organic & biomolecular chemistry, 8(5), 980-983 (2010-02-19)
The multicomponent reaction between proline lithium salt, 2-cyclohexen-1-one and aliphatic aldehydes affords the 4-alkylidene-2-cyclohexen-1-ones, which are interesting fragrances, and bicyclic amino acids that bear four additional stereocenters, obtained as single stereoisomer.

Our team of scientists has experience in all areas of research including Life Science, Material Science, Chemical Synthesis, Chromatography, Analytical and many others.

Contact Technical Service