Skip to Content
Merck
All Photos(1)

Documents

916757

Sigma-Aldrich

Stainless steel dispensing tips

21G, 0.50″, 50/box

Synonym(s):

3d bioprinting consumables, Blunt needle

Sign Into View Organizational & Contract Pricing


About This Item

UNSPSC Code:
42182011
NACRES:
NB.51

Quality Level

description

Stainless steel dispensing tips, 21 Gauge, 0.50″, purple, 50/box

Application

Stainless steel dispensing tips are 21 gauge blunt needles.The tips are free of flash, burrs, and contaminants.

Certificates of Analysis (COA)

Search for Certificates of Analysis (COA) by entering the products Lot/Batch Number. Lot and Batch Numbers can be found on a product’s label following the words ‘Lot’ or ‘Batch’.

Already Own This Product?

Find documentation for the products that you have recently purchased in the Document Library.

Visit the Document Library

Our team of scientists has experience in all areas of research including Life Science, Material Science, Chemical Synthesis, Chromatography, Analytical and many others.

Contact Technical Service