Skip to Content
Merck
All Photos(1)

Key Documents

908320

Sigma-Aldrich

2-Methylglutaric-4,5-13C2 acid solution

1 mg/mL in methanol, ≥98 atom % 13C, ≥95% (CP)

Sign Into View Organizational & Contract Pricing


About This Item

Empirical Formula (Hill Notation):
13C2C4H10O4
Molecular Weight:
148.13
UNSPSC Code:
12352106
NACRES:
NA.12

isotopic purity

≥98 atom % 13C

Quality Level

Assay

≥95% (CP)

form

solution

concentration

1 mg/mL in methanol

technique(s)

mass spectrometry (MS): suitable

mass shift

M+2

storage temp.

−20°C

SMILES string

O[13C]([13CH2]CC(C)C(O)=O)=O

Looking for similar products? Visit Product Comparison Guide

Packaging

This product may be available from bulk stock and can be packaged on demand. For information on pricing, availability and packaging, please contact Stable Isotopes Customer Service.

Signal Word

Danger

Hazard Classifications

Acute Tox. 3 Dermal - Acute Tox. 3 Inhalation - Acute Tox. 3 Oral - Flam. Liq. 2 - STOT SE 1

Target Organs

Eyes,Central nervous system

Storage Class Code

3 - Flammable liquids

WGK

WGK 2

Flash Point(F)

49.5 °F

Flash Point(C)

9.7 °C


Choose from one of the most recent versions:

Certificates of Analysis (COA)

Lot/Batch Number

Sorry, we don't have COAs for this product available online at this time.

If you need assistance, please contact Customer Support.

Already Own This Product?

Find documentation for the products that you have recently purchased in the Document Library.

Visit the Document Library

Our team of scientists has experience in all areas of research including Life Science, Material Science, Chemical Synthesis, Chromatography, Analytical and many others.

Contact Technical Service