Skip to Content
Merck
All Photos(3)

Key Documents

901452

Sigma-Aldrich

Guanidinium bromide

greener alternative

≥99%, anhydrous

Sign Into View Organizational & Contract Pricing


About This Item

Empirical Formula (Hill Notation):
CH6BrN3
CAS Number:
Molecular Weight:
139.98
MDL number:
UNSPSC Code:
12352200
NACRES:
NA.23

grade

anhydrous

Assay

≥99%

form

powder

greener alternative product characteristics

Design for Energy Efficiency
Learn more about the Principles of Green Chemistry.

sustainability

Greener Alternative Product

greener alternative category

SMILES string

NC(N)=N.[H]Br

InChI

1S/CH5N3.BrH/c2-1(3)4;/h(H5,2,3,4);1H

InChI key

VQNVZLDDLJBKNS-UHFFFAOYSA-N

Looking for similar products? Visit Product Comparison Guide

Application

Guanidinium bromide (GBr) can be used in the functionalization of poly(2,6-dimethyl-1,4-phenylene oxide) (PPO) for the formation of the anion exchange membrane for fuel cells applications. It can also be used in the preparation of an amphiphilic catalyst for the production of biodiesel.
Organohalide based perovskites have emerged as an important class of material for solar cell applications. Our perovskites precursors with extremely low water contents are useful for synthesizing mixed cation or anion perovskites needed for the optimization of the band gap, carrier diffusion length and power conversion efficiency of perovskites based solar cells.

Caution

  • Extremely hygroscopic.
  • Handle in glove box.
  • Handle and store under nitrogen atmosphere.

Storage Class Code

11 - Combustible Solids

WGK

WGK 3

Flash Point(F)

Not applicable

Flash Point(C)

Not applicable


Choose from one of the most recent versions:

Certificates of Analysis (COA)

Lot/Batch Number

It looks like we've run into a problem, but you can still download Certificates of Analysis from our Documents section.

If you need assistance, please contact Customer Support.

Already Own This Product?

Find documentation for the products that you have recently purchased in the Document Library.

Visit the Document Library

A facile strategy for the synthesis of guanidinium-functionalized polymer as alkaline anion exchange membrane with improved alkaline stability
Liu L, et al.
Journal of Membrane Science , 453, 52-60 (2014)
Recent Advances in Hybrid Halide Perovskites-based Solar Cells.
Kalyanasundaram K, et al.
Material Matters, 11, 3-3 (2016)
Catalytic performance of a novel amphiphilic alkaline ionic liquid for biodiesel production: influence of basicity and conductivity
Zhang P, et al.
Renewable Energy, 86, 99-105 (2016)
Entropic stabilization of mixed A-cation ABX3 metal halide perovskites for high performance perovskite solar cells.
Yi C, et al.
Energy & Environmental Science, 9, 656-656 (2016)
Recent Advances in Hybrid Halide Perovskites-based Solar Cells.
Kalyanasundaram K, et al.
Material Matters, 11, 3-3 (2016)

Articles

Next generation solar cells have the potential to achieve conversion efficiencies beyond the Shockley-Queisser (S-Q) limit while also significantly lowering production costs.

Dr. Perini and Professor Correa-Baena discuss the latest research and effort to obtain higher performance and stability of perovskite materials.

Our team of scientists has experience in all areas of research including Life Science, Material Science, Chemical Synthesis, Chromatography, Analytical and many others.

Contact Technical Service