901062
Pinacol (dichloromethyl) boronate
≥95%
Synonym(s):
1,3,2-Dioxaborolane, 2-(dichloromethyl)-4,4,5,5-tetramethyl-1,3,2-dioxaborolane, Dichloromethyl pinacol boronate
Sign Into View Organizational & Contract Pricing
Select a Size
1 G
CZK 3,675.00
5 G
CZK 17,300.00
CZK 3,675.00
List PriceCZK 5,250.00Save 30%Please contact Customer Service for Availability
Request a Bulk Order
All Photos(1)
Select a Size
Change View
1 G
CZK 3,675.00
5 G
CZK 17,300.00
About This Item
Empirical Formula (Hill Notation):
C7H13BCl2O2
CAS Number:
Molecular Weight:
210.89
MDL number:
UNSPSC Code:
12352200
NACRES:
NA.22
CZK 3,675.00
List PriceCZK 5,250.00Save 30%Please contact Customer Service for Availability
Request a Bulk Order
Quality Level
Assay
≥95%
form
(Powder or Solid or Crystals or Semi-Solid or Liquid)
refractive index
n/D 1.4511
density
1.1423
functional group
chloro
SMILES string
B1(OC(C(O1)(C)C)(C)C)C(Cl)Cl
InChI
1S/C7H13BCl2O2/c1-6(2)7(3,4)12-8(11-6)5(9)10/h5H,1-4H3
InChI key
GQFQFYFLABSDDS-UHFFFAOYSA-N
Related Categories
Application
Pinacol (dichloromethyl) boronate is a general reagent that was employed by Prof. Greg C. Fu and coworkers to achieve the iterative synthesis of asymmetric (secondary) alkyl boronate esters.[1] These can be further derivatized through a growing suite of chemical transformations developed in the last decade (cross coupling, oxidation, amination, halogenation) that proceed with stereochemical fidelity.
Storage Class Code
10 - Combustible liquids
WGK
WGK 3
Flash Point(F)
201.2 °F
Flash Point(C)
94 °C
Choose from one of the most recent versions:
Already Own This Product?
Find documentation for the products that you have recently purchased in the Document Library.
A general, modular method for the catalytic asymmetric synthesis of alkylboronate esters.
Schmidt J, et al.
Science, 354(6317), 1265-1269 (2016)
Active Filters
Our team of scientists has experience in all areas of research including Life Science, Material Science, Chemical Synthesis, Chromatography, Analytical and many others.
Contact Technical Service