Skip to Content
Merck
All Photos(1)

Key Documents

901018

Sigma-Aldrich

Lithium-7Li sulfide

≥99 atom % 7Li, ≥99% (CP)

Sign Into View Organizational & Contract Pricing

Select a Size

1 G
CZK 68,800.00
5 G
CZK 200,600.00

CZK 68,800.00


Please contact Customer Service for Availability


Select a Size

Change View
1 G
CZK 68,800.00
5 G
CZK 200,600.00

About This Item

Empirical Formula (Hill Notation):
7Li2S
Molecular Weight:
46.10
UNSPSC Code:
12352302
NACRES:
NA.12

CZK 68,800.00


Please contact Customer Service for Availability

isotopic purity

≥99 atom % 7Li

Quality Level

Assay

≥99% (CP)

form

solid

mass shift

depleted

SMILES string

[7Li][7Li].S

InChI

1S/2Li.H2S/h;;1H2/i2*1+0;

InChI key

BQMZCPKSFSMEGX-ONPSQTMSSA-N

Related Categories

Packaging

This product may be available from bulk stock and can be packaged on demand. For information on pricing, availability and packaging, please contact Stable Isotopes Customer Service.

Pictograms

Skull and crossbonesCorrosion

Signal Word

Danger

Hazard Statements

Hazard Classifications

Acute Tox. 3 Oral - Skin Corr. 1B

Storage Class Code

6.1A - Combustible acute toxic Cat. 1 and 2 / very toxic hazardous materials

WGK

WGK 3

Flash Point(F)

Not applicable

Flash Point(C)

Not applicable


Choose from one of the most recent versions:

Certificates of Analysis (COA)

Lot/Batch Number

Don't see the Right Version?

If you require a particular version, you can look up a specific certificate by the Lot or Batch number.

Already Own This Product?

Find documentation for the products that you have recently purchased in the Document Library.

Visit the Document Library

Questions

Reviews

No rating value

Active Filters

Our team of scientists has experience in all areas of research including Life Science, Material Science, Chemical Synthesis, Chromatography, Analytical and many others.

Contact Technical Service