Skip to Content
Merck
All Photos(1)

Documents

900857

Sigma-Aldrich

Tributylmethylammonium bis(trifluoromethylsulfonyl)imide

≥99%, H2O <500 ppm

Synonym(s):

Methyltributylammonium bis(trifluoromethylsulfonyl)imide, N 4441-TFSI

Sign Into View Organizational & Contract Pricing


About This Item

Empirical Formula (Hill Notation):
C15H30F6N2O4S2
CAS Number:
Molecular Weight:
480.53
MDL number:
UNSPSC Code:
12352111
NACRES:
NA.23

Assay

≥99%

form

liquid

composition

H2O, <500 ppm

impurities

≤500 ppm H2O

mp

27 °C

density

1.276 g/cm3

application(s)

battery manufacturing

InChI

1S/C13H30N.C2F6NO4S2/c1-5-8-11-14(4,12-9-6-2)13-10-7-3;3-1(4,5)14(10,11)9-15(12,13)2(6,7)8/h5-13H2,1-4H3;/q+1;-1

InChI key

XALVHDZWUBSWES-UHFFFAOYSA-N

General description

Tributylmethylammonium bis(trifluoromethylsulfonyl)imide is a class of electrolytic materials that can be used in the fabrication of lithium-ion batteries. Lithium-ion batteries consist of anode, cathode, and electrolyte with a charge-discharge cycle. These materials enable the formation of greener and sustainable batteries for electrical energy storage.

Application

Ionic liquids (ILs) are molten salts with melting points lower than 100 °C. They usually consist of pair of organic cation and anion. ILs exhibit unique properties such as non-volatility, high thermal stability, and high ionic conductivity and find applications as electrolytes in lithium/sodium ion batteries and dye-sensitized solar cells. They are also used as media for synthesis of conducting polymers and intercalation electrode materials.

Storage Class Code

10 - Combustible liquids

WGK

WGK 3

Flash Point(F)

230.0 °F

Flash Point(C)

110 °C


Certificates of Analysis (COA)

Search for Certificates of Analysis (COA) by entering the products Lot/Batch Number. Lot and Batch Numbers can be found on a product’s label following the words ‘Lot’ or ‘Batch’.

Already Own This Product?

Find documentation for the products that you have recently purchased in the Document Library.

Visit the Document Library

Ionic liquids and their solid-state analogues as materials for energy generation and storage.
MacFarlane DR, et al.
Nature Reviews. Materials, 1, 15005-15005 (2016)
Masayoshi Watanabe et al.
Chemical reviews, 117(10), 7190-7239 (2017-01-14)
Ionic liquids (ILs) are liquids consisting entirely of ions and can be further defined as molten salts having melting points lower than 100 °C. One of the most important research areas for IL utilization is undoubtedly their energy application, especially
Dandan Han et al.
Molecules (Basel, Switzerland), 15(4), 2405-2426 (2010-04-30)
Ionic liquids (ILs) have been applied in different areas of separation, such as ionic liquid supported membranes, as mobile phase additives and surface-bonded stationary phases in chromatography separations and as the extraction solvent in sample preparations, because they can be
Challenges for rechargeable Li batteries
Goodenough JB and Kim Y
Chemistry of Materials, 22(3), 587-603 (2009)
Electrodes with high power and high capacity for rechargeable lithium batteries
Kang K, et al.
Science, 311(5763), 977-980 (2006)

Articles

Solid-state Li batteries: Review of solid electrolytes, ion conduction, structures, and electrochemical processes.

Ionic liquid electrolytes explored for rechargeable batteries' advancement; future IL development discussed.

Our team of scientists has experience in all areas of research including Life Science, Material Science, Chemical Synthesis, Chromatography, Analytical and many others.

Contact Technical Service