Skip to Content
Merck
All Photos(2)

Documents

900320

Sigma-Aldrich

Dyed poly(dioxanone)

viscosity 1.5 dL/g 

Sign Into View Organizational & Contract Pricing


About This Item

Empirical Formula (Hill Notation):
(C4H6O3)n
CAS Number:
UNSPSC Code:
41116100

form

chips or granules

viscosity

1.5 dL/g

storage temp.

−20°C

InChI

1S/C4H6O3/c5-4-3-6-1-2-7-4/h1-3H2

InChI key

VPVXHAANQNHFSF-UHFFFAOYSA-N

Looking for similar products? Visit Product Comparison Guide

Storage Class Code

11 - Combustible Solids

WGK

WGK 3

Flash Point(F)

Not applicable

Flash Point(C)

Not applicable


Certificates of Analysis (COA)

Search for Certificates of Analysis (COA) by entering the products Lot/Batch Number. Lot and Batch Numbers can be found on a product’s label following the words ‘Lot’ or ‘Batch’.

Already Own This Product?

Find documentation for the products that you have recently purchased in the Document Library.

Visit the Document Library

Articles

Main features of p-dioxanone, glycolide and lactide based copolymers

Innovations in polymer technology have had a significant impact on the advancement of novel drug delivery systems.

Our team of scientists has experience in all areas of research including Life Science, Material Science, Chemical Synthesis, Chromatography, Analytical and many others.

Contact Technical Service