Skip to Content
Merck
All Photos(2)

Documents

900001

Sigma-Aldrich

tert-Amylbenzene

≥99%, H2O <100 ppm, acid <200 ppm

Synonym(s):

2-Methyl-2-phenylbutane, tert-Pentylbenzene

Sign Into View Organizational & Contract Pricing


About This Item

Empirical Formula (Hill Notation):
C11H16
CAS Number:
Molecular Weight:
148.24
MDL number:
UNSPSC Code:
26111700
PubChem Substance ID:

grade

battery grade

Quality Level

Assay

≥99%

form

liquid

composition

acid, <200 ppm
H2O, <100 ppm

impurities

≤100 ppm H2O
≤200 ppm acid

application(s)

battery manufacturing

SMILES string

CCC(C)(C)C1=CC=CC=C1

InChI

1S/C11H16/c1-4-11(2,3)10-8-6-5-7-9-10/h5-9H,4H2,1-3H3

InChI key

QHTJSSMHBLGUHV-UHFFFAOYSA-N

Looking for similar products? Visit Product Comparison Guide

Application

The presence of electrolyte additives enhances the performance of lithium ion batteries by improving their thermal stability and ionic conductivity. Additives also reduce the flammability of electrolytes and provide overcharge protection. t-Amyl benzene finds application as additive in lithium ion batteries and its addition in grade would be good expansion of our LIB portfolio.

Legal Information

Product of MU Ionic Solutions Corp

Signal Word

Warning

Hazard Statements

Hazard Classifications

Aquatic Chronic 2 - Flam. Liq. 3 - Skin Sens. 1

Storage Class Code

3 - Flammable liquids

WGK

WGK 3

Flash Point(F)

136.4 °F

Flash Point(C)

58 °C


Certificates of Analysis (COA)

Search for Certificates of Analysis (COA) by entering the products Lot/Batch Number. Lot and Batch Numbers can be found on a product’s label following the words ‘Lot’ or ‘Batch’.

Already Own This Product?

Find documentation for the products that you have recently purchased in the Document Library.

Visit the Document Library

Zhang S.
Journal of Power Sources, 162, 1379-1379 (2006)

Articles

Experts discuss challenges and production processes of nickel-rich layered oxide cathode materials in energy storage systems.

Our team of scientists has experience in all areas of research including Life Science, Material Science, Chemical Synthesis, Chromatography, Analytical and many others.

Contact Technical Service