Skip to Content
Merck
All Photos(1)

Documents

87250

Sigma-Aldrich

Tetradodecylammonium chloride

≥97.0% (AT)

Sign Into View Organizational & Contract Pricing


About This Item

Linear Formula:
[CH3(CH2)10CH2]4N(Cl)
CAS Number:
Molecular Weight:
726.77
Beilstein:
8536098
MDL number:
UNSPSC Code:
12352116
PubChem Substance ID:
NACRES:
NA.22

Assay

≥97.0% (AT)

form

crystals

mp

81-84 °C

SMILES string

[Cl-].CCCCCCCCCCCC[N+](CCCCCCCCCCCC)(CCCCCCCCCCCC)CCCCCCCCCCCC

InChI

1S/C48H100N.ClH/c1-5-9-13-17-21-25-29-33-37-41-45-49(46-42-38-34-30-26-22-18-14-10-6-2,47-43-39-35-31-27-23-19-15-11-7-3)48-44-40-36-32-28-24-20-16-12-8-4;/h5-48H2,1-4H3;1H/q+1;/p-1

InChI key

KEVJUENWYCMNET-UHFFFAOYSA-M

Pictograms

Exclamation mark

Signal Word

Warning

Hazard Statements

Hazard Classifications

Eye Irrit. 2 - Skin Irrit. 2 - STOT SE 3

Target Organs

Respiratory system

Storage Class Code

11 - Combustible Solids

WGK

WGK 3

Flash Point(F)

Not applicable

Flash Point(C)

Not applicable

Personal Protective Equipment

dust mask type N95 (US), Eyeshields, Gloves

Certificates of Analysis (COA)

Search for Certificates of Analysis (COA) by entering the products Lot/Batch Number. Lot and Batch Numbers can be found on a product’s label following the words ‘Lot’ or ‘Batch’.

Already Own This Product?

Find documentation for the products that you have recently purchased in the Document Library.

Visit the Document Library

Ernő Lindner et al.
Membranes, 10(9) (2020-09-02)
In the last 50 years, plasticized polyvinyl chloride (PVC) membranes have gained unique importance in chemical sensor development. Originally, these membranes separated two solutions in conventional ion-selective electrodes. Later, the same membranes were applied over a variety of supporting electrodes
Eduardo Laborda et al.
Angewandte Chemie (International ed. in English), 56(3), 782-785 (2016-12-10)
A new electrochemical framework for tracking individual soft particles in solution and monitoring their fusion with polarized liquid-liquid interfaces is reported. The physicochemical principle lies in the interfacial transfer of an ionic probe confined in the particles dispersed in solution
James B Sheppard et al.
The Analyst, 142(6), 930-937 (2017-02-22)
The diffusion-controlled transport of ions and molecules through polymer membranes utilized in chemical and biosensors is often the key factor determining the response characteristics of these sensors. In this paper, a simple voltammetric method is described for the determination of
Hao Yu et al.
The journal of physical chemistry. B, 119(29), 9319-9334 (2014-10-08)
The electric potential difference across cell membranes, known as the membrane potential, plays an important role in the activation of many biological processes. To investigate the effect of the membrane potential on the molecular ordering of lipids within a biomimetic

Our team of scientists has experience in all areas of research including Life Science, Material Science, Chemical Synthesis, Chromatography, Analytical and many others.

Contact Technical Service