Skip to Content
Merck
All Photos(1)

Key Documents

86652

Sigma-Aldrich

Tetraethylammonium (meta)periodate

technical, moistened with water, ≥90% (calc. based on dry substance, RT)

Synonym(s):

N,N,N-Triethylethanaminium salt with periodic acid

Sign Into View Organizational & Contract Pricing


About This Item

Linear Formula:
(C2H5)4N(IO4)
CAS Number:
Molecular Weight:
321.15
Beilstein:
4166132
EC Number:
MDL number:
UNSPSC Code:
12352000
PubChem Substance ID:
NACRES:
NA.22

grade

technical

Assay

≥90% (calc. based on dry substance, RT)

form

crystals

quality

moistened with water

contains

~10% water as stabilizer

reaction suitability

reagent type: oxidant

mp

~170 °C (dec.)

SMILES string

[O-]I(=O)(=O)=O.CC[N+](CC)(CC)CC

InChI

1S/C8H20N.HIO4/c1-5-9(6-2,7-3)8-4;2-1(3,4)5/h5-8H2,1-4H3;(H,2,3,4,5)/q+1;/p-1

InChI key

YXYPONURGKGPDV-UHFFFAOYSA-M

Application

Reactant for intramolecular cycloaddition reactions
Tetraethylammonium (meta)periodate can be used as an oxidant in the N-dealkylation of tertiary amines using Fe(III) complex as a catalyst. It can also be employed as a reagent in the intramolecular cycloaddition reactions.

Other Notes

Oxidant for use in organic solvents; Oxidation of 1,2-diols and olefins

Pictograms

Flame

Signal Word

Danger

Hazard Statements

Precautionary Statements

Hazard Classifications

Flam. Sol. 1

Storage Class Code

4.1B - Flammable solid hazardous materials

WGK

WGK 3

Flash Point(F)

Not applicable

Flash Point(C)

Not applicable

Personal Protective Equipment

dust mask type N95 (US), Eyeshields, Gloves

Choose from one of the most recent versions:

Certificates of Analysis (COA)

Lot/Batch Number

Don't see the Right Version?

If you require a particular version, you can look up a specific certificate by the Lot or Batch number.

Already Own This Product?

Find documentation for the products that you have recently purchased in the Document Library.

Visit the Document Library

Oxidative N-Dealkylation of Tertiary Amines with Tetraethylammonium Periodate Catalyzed by Metal Complexes
Bhat, Daisy and Sharma, Nidhi
Australian Journal of Chemistry, 70(3), 233-236 (2017)
G.E. Keck
Tetrahedron Letters, 4767-4767 (1978)
A.A. Freer et al.
Journal of the Chemical Society. Perkin Transactions 1, 1001-1001 (1991)
Synthesis of (2R, 5R)-2-amino-5-hydroxyhexanoic acid by intramolecular cycloaddition
Sheradsky T and Silcoff ER
Molecules (Basel), 3(3), 80-87 (1998)
K. Inomata et al.
Bulletin of the Chemical Society of Japan, 53, 565-565 (1980)

Our team of scientists has experience in all areas of research including Life Science, Material Science, Chemical Synthesis, Chromatography, Analytical and many others.

Contact Technical Service