Skip to Content
Merck
All Photos(1)

Key Documents

857777

Sigma-Aldrich

Cytochalasin B from Helminthosporium dematioideum

Sign Into View Organizational & Contract Pricing


About This Item

Empirical Formula (Hill Notation):
C29H37NO5
CAS Number:
Molecular Weight:
479.61
Beilstein:
1096207
EC Number:
MDL number:
UNSPSC Code:
12352103
PubChem Substance ID:

Quality Level

SMILES string

C[C@@H]1CCC[C@@H](O)\C=C\C(=O)O[C@]23[C@@H](\C=C\C1)[C@H](O)C(=C)[C@@H](C)[C@H]2[C@H](Cc4ccccc4)NC3=O

InChI

1S/C29H37NO5/c1-18-9-7-13-22(31)15-16-25(32)35-29-23(14-8-10-18)27(33)20(3)19(2)26(29)24(30-28(29)34)17-21-11-5-4-6-12-21/h4-6,8,11-12,14-16,18-19,22-24,26-27,31,33H,3,7,9-10,13,17H2,1-2H3,(H,30,34)/b14-8+,16-15+/t18-,19-,22-,23+,24+,26+,27-,29-/m1/s1

InChI key

GBOGMAARMMDZGR-TYHYBEHESA-N

Pictograms

Skull and crossbonesHealth hazard

Signal Word

Danger

Hazard Statements

Hazard Classifications

Acute Tox. 1 Inhalation - Acute Tox. 2 Dermal - Acute Tox. 2 Oral - Repr. 2

Storage Class Code

6.1A - Combustible acute toxic Cat. 1 and 2 / very toxic hazardous materials

WGK

WGK 3

Personal Protective Equipment

dust mask type N95 (US), Eyeshields, Gloves

Choose from one of the most recent versions:

Certificates of Analysis (COA)

Lot/Batch Number

Don't see the Right Version?

If you require a particular version, you can look up a specific certificate by the Lot or Batch number.

Already Own This Product?

Find documentation for the products that you have recently purchased in the Document Library.

Visit the Document Library

F Marco-Jiménez et al.
Cryo letters, 33(1), 24-30 (2012-03-22)
The study tested the efficacy of pre-treating mature porcine oocytes with cytochalasin B before vitrification by the open pulled straw method (OPS) in a low toxicity solution containing ice blockers. The effects of pre-treating the oocytes with 7.5 micrograms per
François Perreault et al.
The Science of the total environment, 441, 117-124 (2012-11-10)
Copper oxide nanoparticles (CuO NPs) are used for their biocide potential however they were also shown to be highly toxic to mammalian cells. Therefore, the effects of CuO NPs should be carefully investigated to determine the most sensitive processes for
Nicolas Vidal et al.
Biochimie, 94(12), 2569-2576 (2012-07-28)
We have developed a rapid nonradioisotope chemiluminescent assay adapted to high-throughput screening experiments, to evaluate glucose uptake activity in cultured cells. For chemiluminescence quantification of 2-deoxyglucose, we used a luminol oxidation reaction after an enzymatic dephosphorylation of 2-deoxyglucose-6-phosphate. All reactions
M A Pogorelova et al.
Ontogenez, 43(2), 94-102 (2012-06-02)
Mouse single-cell embryos exhibit robust Regulatory Volume Decrease (RVD). In what manner the very early mammalian embryo following zygote stage is appreciably altered by the anisotonic extracellular solution is, as yet, totally unclear. Little attention was paid to this direction
C Milligan et al.
The Journal of physiology, 591(6), 1463-1474 (2013-01-09)
Abstract  Red blood cells (RBCs) from patients with sickle cell disease (SCD) lyse in deoxygenated isosmotic non-electrolyte solutions. Haemolysis has features which suggest that it is linked to activation of the pathway termed Psickle. This pathway is usually described as

Our team of scientists has experience in all areas of research including Life Science, Material Science, Chemical Synthesis, Chromatography, Analytical and many others.

Contact Technical Service