Skip to Content
Merck
All Photos(1)

Key Documents

856770

Sigma-Aldrich

DL-Alanine β-naphthylamide hydrochloride

98%

Sign Into View Organizational & Contract Pricing


About This Item

Linear Formula:
CH3CH(NH2)CONHC10H7·HCl
CAS Number:
Molecular Weight:
250.72
EC Number:
MDL number:
UNSPSC Code:
12352103

Assay

98%

mp

258-260 °C (dec.) (lit.)

SMILES string

Cl.CC(N)C(=O)Nc1ccc2ccccc2c1

InChI

1S/C13H14N2O.ClH/c1-9(14)13(16)15-12-7-6-10-4-2-3-5-11(10)8-12;/h2-9H,14H2,1H3,(H,15,16);1H

InChI key

WNLRRMRLNYQNOZ-UHFFFAOYSA-N

Looking for similar products? Visit Product Comparison Guide

replaced by

Product No.
Description
Pricing

Choose from one of the most recent versions:

Certificates of Analysis (COA)

Lot/Batch Number

Don't see the Right Version?

If you require a particular version, you can look up a specific certificate by the Lot or Batch number.

Already Own This Product?

Find documentation for the products that you have recently purchased in the Document Library.

Visit the Document Library

Our team of scientists has experience in all areas of research including Life Science, Material Science, Chemical Synthesis, Chromatography, Analytical and many others.

Contact Technical Service