Skip to Content
Merck
All Photos(1)

Key Documents

84738

Sigma-Aldrich

Sulfur trioxide pyridine complex polymer-bound

extent of labeling: ≥2.0 mmol/g SO3 loading

Synonym(s):

Poly(4-vinylpyridine) sulfur trioxide complex, Pyridine sulfur trioxide complex polymer-bound

Sign Into View Organizational & Contract Pricing


About This Item

UNSPSC Code:
12352200
PubChem Substance ID:

extent of labeling

≥2.0 mmol/g SO3 loading

SMILES string

O=S(=O)=O.c1ccncc1

Other Notes

Polymer-supported reagent which can be used analogously to the monomeric reagent 84737 above. The consumed reagent can be simply removed by filtration

Storage Class Code

13 - Non Combustible Solids

WGK

WGK 3

Flash Point(F)

Not applicable

Flash Point(C)

Not applicable

Personal Protective Equipment

dust mask type N95 (US), Eyeshields, Gloves

Certificates of Analysis (COA)

Search for Certificates of Analysis (COA) by entering the products Lot/Batch Number. Lot and Batch Numbers can be found on a product’s label following the words ‘Lot’ or ‘Batch’.

Already Own This Product?

Find documentation for the products that you have recently purchased in the Document Library.

Visit the Document Library

W. Graf
Chemistry & Industry, 232-232 (1987)

Our team of scientists has experience in all areas of research including Life Science, Material Science, Chemical Synthesis, Chromatography, Analytical and many others.

Contact Technical Service