Skip to Content
Merck
All Photos(1)

Key Documents

82366

Sigma-Aldrich

3-(tert-Butoxy)propylamine

technical, ≥90% (GC)

Synonym(s):

3-Aminopropyl tert-butyl ether

Sign Into View Organizational & Contract Pricing


About This Item

Linear Formula:
(H3C)3COCH2CH2CH2NH2
CAS Number:
Molecular Weight:
131.22
MDL number:
UNSPSC Code:
12352100
PubChem Substance ID:

grade

technical

Assay

≥90% (GC)

SMILES string

CC(C)(C)OCCCN

Pictograms

Corrosion

Signal Word

Danger

Hazard Statements

Hazard Classifications

Skin Corr. 1B

Storage Class Code

8A - Combustible corrosive hazardous materials

WGK

WGK 3

Flash Point(F)

Not applicable

Flash Point(C)

Not applicable

Personal Protective Equipment

dust mask type N95 (US), Eyeshields, Gloves

Choose from one of the most recent versions:

Certificates of Analysis (COA)

Lot/Batch Number

Don't see the Right Version?

If you require a particular version, you can look up a specific certificate by the Lot or Batch number.

Already Own This Product?

Find documentation for the products that you have recently purchased in the Document Library.

Visit the Document Library

Our team of scientists has experience in all areas of research including Life Science, Material Science, Chemical Synthesis, Chromatography, Analytical and many others.

Contact Technical Service