Skip to Content
Merck
All Photos(1)

Documents

82356

Sigma-Aldrich

3,5,5-Trimethylhexyl 3,5,5-trimethylhexanoate

technical, ≥70% (GC)

Synonym(s):

3,5,5-Trimethylhexyl 3,5,5-trimethylcaproat

Sign Into View Organizational & Contract Pricing


About This Item

Linear Formula:
(CH3)3CCH2CH(CH3)CH2COOCH2CH2CH(CH)3CH2C(CH3)3
CAS Number:
Molecular Weight:
284.48
Beilstein:
1785018
EC Number:
MDL number:
UNSPSC Code:
12352100
PubChem Substance ID:

grade

technical

Assay

≥70% (GC)

refractive index

n20/D 1.437

acid value

≤1.0

iodine value

≤1.0

saponification value

185‑200

transition temp

solidification point ≤−15 °C

density

0.856 g/mL at 20 °C (lit.)

SMILES string

CC(CCOC(=O)CC(C)CC(C)(C)C)CC(C)(C)C


Certificates of Analysis (COA)

Search for Certificates of Analysis (COA) by entering the products Lot/Batch Number. Lot and Batch Numbers can be found on a product’s label following the words ‘Lot’ or ‘Batch’.

Already Own This Product?

Find documentation for the products that you have recently purchased in the Document Library.

Visit the Document Library

Our team of scientists has experience in all areas of research including Life Science, Material Science, Chemical Synthesis, Chromatography, Analytical and many others.

Contact Technical Service