Sign In to View Organizational & Contract Pricing.
Select a Size
Change View
About This Item
Empirical Formula (Hill Notation):
C25H33F4NO5
CAS Number:
Molecular Weight:
503.53
UNSPSC Code:
12352200
PubChem Substance ID:
NACRES:
NA.22
Form:
powder
form
powder
storage temp.
2-8°C
SMILES string
FC1=C(C(C)=O)C(F)=C(F)C(C2=C([O-])OC3(CCCCC3)OC2=O)=C1F.CC[NH+](C(C)C)C(C)C
InChI
1S/C17H14F4O5.C8H19N/c1-7(22)8-11(18)13(20)9(14(21)12(8)19)10-15(23)25-17(26-16(10)24)5-3-2-4-6-17;1-6-9(7(2)3)8(4)5/h23H,2-6H2,1H3;7-8H,6H2,1-5H3
InChI key
UYTIIFFVFGALQP-UHFFFAOYSA-N
General description
N-Ethyl-N-isopropylpropan-2-aminium 3-(4-acetyl-2,3,5,6-tetrafluorophenyl)-4-oxo-1,5-dioxaspiro[5.5]-undec-2-en-2-olate is a fluoroarylated-Meldrum′s acid adduct. It can be prepared by reacting 2′,3′,4′,5′,6′-pentafluoroacetophenone with cyclohexyl-Meldrum′s acid in the presence of N,N-diisopropylethylamine and acetonitrile.
Application
This compound undergoes cycloreversion to form the fluoro(hetero)aryl ketene, which undergoes efficient coupling with nucleophiles and allows rapid incorporation of highly fluorinated α-fluoro(hetero)aryl acetic acid derivatives.
Choose from one of the most recent versions:
Already Own This Product?
Find documentation for the products that you have recently purchased in the Document Library.