Skip to Content
Merck
All Photos(3)

Key Documents

807532

Sigma-Aldrich

Silver nanoplates

resonant wavelength, 550 nm, ~1 mg/mL (in water with 5 mM sodium borate buffer), PVP functionalized

Sign Into View Organizational & Contract Pricing


About This Item

UNSPSC Code:
12352200
NACRES:
NA.23

form

nanoplates

concentration

~1 mg/mL (in water with 5 mM sodium borate buffer)

resonant wavelength

550 nm

functional group

PVP

storage temp.

2-8°C

Looking for similar products? Visit Product Comparison Guide

Application

Silver nanomaterials have unique physical, chemical and optical properties that are currently being leveraged for a wide variety of biological applications [1-9]. A resurgence of interest in the utility of silver as a broad based antimicrobial agent has led to the development of hundreds of products that incorporate silver nanoparticles to prevent bacterial growth on surfaces [1-4]. Additionally, silver nanoparticles have an optical color that is a function of the silver nanoparticle′s size and shape. The strong coupling of silver nanoparticles to specific wavelengths of incident light can be utilized to develop ultrabright reporter molecules [5-7], highly efficient thermal absorbers, and nanoscale “antennas” that can amplify the strength of the local electromagnetic field

Pictograms

Environment

Signal Word

Warning

Hazard Statements

Precautionary Statements

Hazard Classifications

Aquatic Acute 1 - Aquatic Chronic 1

Storage Class Code

12 - Non Combustible Liquids

WGK

WGK 3

Flash Point(F)

Not applicable

Flash Point(C)

Not applicable


Regulatory Listings

Regulatory Listings are mainly provided for chemical products. Only limited information can be provided here for non-chemical products. No entry means none of the components are listed. It is the user’s obligation to ensure the safe and legal use of the product.

EU REACH SVHC Candidate List

CAS No.

Choose from one of the most recent versions:

Certificates of Analysis (COA)

Lot/Batch Number

It looks like we've run into a problem, but you can still download Certificates of Analysis from our Documents section.

If you need assistance, please contact Customer Support.

Already Own This Product?

Find documentation for the products that you have recently purchased in the Document Library.

Visit the Document Library

Our team of scientists has experience in all areas of research including Life Science, Material Science, Chemical Synthesis, Chromatography, Analytical and many others.

Contact Technical Service