Skip to Content
Merck
All Photos(1)

Documents

802603

Sigma-Aldrich

Aquivion® D83-06A

liquid, dispersion, 6% polymer content in lower aliphatic alcohols and water, water 20 %

Synonym(s):

Aquivion® SO3H-form, Tetrafluoroethylene-perfluoro(3-oxa-4-pentenesulfonic acid) copolymer, Ethanesulfonic acid

Sign Into View Organizational & Contract Pricing


About This Item

Linear Formula:
(C4HF7O4S .C2F4)x
UNSPSC Code:
26111700
NACRES:
NA.23

form

dispersion
liquid

eq. wt.

790±20g/mole (SO3H)

composition

1-propanol, 40 wt. %
2-propanol, 40 wt. %
water, 20%

concentration

6±0.5 % (w/w)

Looking for similar products? Visit Product Comparison Guide

General description

Aquivion® PFSA dispersions are based on the short-side-chain (SSC) copolymer of Tetrafluoroethylene and the Sulfonyl Fluoride Vinyl Ether (SFVE) CF2=CF2-O-(CF2)2-SO2F industrially produced by Solvay Specialty Polymers. The resulting perfluoropolymer′s functional groups are in their sulfonic acid form, SO3H.

Application

Amongst other, Aquivion® D83-24B can be used as formulation additive for production of polymer electrolyte fuel cell electrodes, manufacture or repair of cation-exchange membranes, surface coatings of PTFE membranes to improve their wetting behavior or for the development of superacid catalysts. Addition of further film-forming and/or other volatile or non-volatile chemicals is necessary to impart the desired processing and end-product characteristics.

Legal Information

Aquivion is a registered trademark of Syensqo Group

Signal Word

Danger

Hazard Statements

Hazard Classifications

Eye Dam. 1 - Flam. Liq. 2 - STOT SE 3

Target Organs

Central nervous system

Storage Class Code

3 - Flammable liquids

WGK

WGK 2

Flash Point(F)

53.6 °F - closed cup

Flash Point(C)

12.0 °C - closed cup


Certificates of Analysis (COA)

Search for Certificates of Analysis (COA) by entering the products Lot/Batch Number. Lot and Batch Numbers can be found on a product’s label following the words ‘Lot’ or ‘Batch’.

Already Own This Product?

Find documentation for the products that you have recently purchased in the Document Library.

Visit the Document Library

Articles

Advances in electrochemical water conversion and understanding PEMFC degradation drive progress in hydrogen technologies.

Our team of scientists has experience in all areas of research including Life Science, Material Science, Chemical Synthesis, Chromatography, Analytical and many others.

Contact Technical Service