Skip to Content
Merck
All Photos(1)

Documents

797162

Sigma-Aldrich

tert-Butyl 8-hydroxy-6-thia-2-azaspiro[3.4]octane-2-carboxylate 6,6-dioxide

95%

Synonym(s):

2-Boc-8-hydroxy-6-thia-2-azaspiro[3.4]octane 6,6-dioxide

Sign Into View Organizational & Contract Pricing


About This Item

Empirical Formula (Hill Notation):
C11H19NO5S
CAS Number:
Molecular Weight:
277.34
MDL number:
UNSPSC Code:
12352100
PubChem Substance ID:
NACRES:
NA.22

Assay

95%

form

powder

storage temp.

2-8°C

SMILES string

OC(CS(C1)(=O)=O)C21CN(C(OC(C)(C)C)=O)C2

InChI

1S/C11H19NO5S/c1-10(2,3)17-9(14)12-5-11(6-12)7-18(15,16)4-8(11)13/h8,13H,4-7H2,1-3H3

InChI key

MLPBVLQLVWQEJM-UHFFFAOYSA-N

Application

Spirocyclic building block was first synthesized by Carreira and coworkers, which provides a new area of chemical space with straightforward functional handles for further diversification.

Pictograms

Skull and crossbones

Signal Word

Danger

Hazard Statements

Hazard Classifications

Acute Tox. 3 Oral

Storage Class Code

6.1C - Combustible acute toxic Cat.3 / toxic compounds or compounds which causing chronic effects

WGK

WGK 3

Flash Point(F)

Not applicable

Flash Point(C)

Not applicable


Certificates of Analysis (COA)

Search for Certificates of Analysis (COA) by entering the products Lot/Batch Number. Lot and Batch Numbers can be found on a product’s label following the words ‘Lot’ or ‘Batch’.

Already Own This Product?

Find documentation for the products that you have recently purchased in the Document Library.

Visit the Document Library

Our team of scientists has experience in all areas of research including Life Science, Material Science, Chemical Synthesis, Chromatography, Analytical and many others.

Contact Technical Service