All Photos(3)
About This Item
Empirical Formula (Hill Notation):
C12Br2F8
CAS Number:
Molecular Weight:
455.92
MDL number:
UNSPSC Code:
12352100
PubChem Substance ID:
NACRES:
NA.22
Recommended Products
Assay
97%
form
crystals
mp
95-100 °C
functional group
bromo
fluoro
SMILES string
FC1=C(F)C(C2=C(F)C(F)=C(F)C(F)=C2Br)=C(Br)C(F)=C1F
InChI
1S/C12Br2F8/c13-3-1(5(15)9(19)11(21)7(3)17)2-4(14)8(18)12(22)10(20)6(2)16
InChI key
CZEFBGCEVOAASZ-UHFFFAOYSA-N
Application
This material is a precursor in the synthesis of Donor-Acceptor systems useful for use as electron transporting materials in blends with P3HT in organic photovoltaic devices.
Signal Word
Warning
Hazard Statements
Precautionary Statements
Hazard Classifications
Eye Irrit. 2 - Skin Irrit. 2 - STOT SE 3
Target Organs
Respiratory system
Storage Class Code
11 - Combustible Solids
WGK
WGK 3
Flash Point(F)
Not applicable
Flash Point(C)
Not applicable
Choose from one of the most recent versions:
Certificates of Analysis (COA)
Lot/Batch Number
Don't see the Right Version?
If you require a particular version, you can look up a specific certificate by the Lot or Batch number.
Already Own This Product?
Find documentation for the products that you have recently purchased in the Document Library.
Katharine Geramita et al.
The Journal of organic chemistry, 75(6), 1871-1887 (2010-03-04)
A series of oligothiophene-perfluoro-9-heterofluorene donor-acceptor (DA) compounds was synthesized via a combination of nucleophilic aromatic substitution (S(N)Ar(F)) and palladium coupling reactions. These compounds are of interest as possible building blocks for materials with useful electron transport properties, since they possess
Our team of scientists has experience in all areas of research including Life Science, Material Science, Chemical Synthesis, Chromatography, Analytical and many others.
Contact Technical Service