Skip to Content
Merck
All Photos(1)

Key Documents

776483

Sigma-Aldrich

8-Acetyl-5,6,7,8-tetrahydroquinoline

95%

Synonym(s):

8-Acetyl-5,6,7,8-tetrahydroquinoline

Sign Into View Organizational & Contract Pricing


About This Item

Empirical Formula (Hill Notation):
C11H13NO
CAS Number:
Molecular Weight:
175.23
MDL number:
UNSPSC Code:
12161600
NACRES:
NA.22

Assay

95%

form

liquid

reaction suitability

reagent type: ligand

refractive index

n20/D 1.568

density

1.107 g/mL at 25 °C

SMILES string

n1c2c(ccc1)CCCC2C(=O)C

InChI

1S/C11H13NO/c1-8(13)10-6-2-4-9-5-3-7-12-11(9)10/h3,5,7,10H,2,4,6H2,1H3

InChI key

YCPXJTSSSPGZOE-UHFFFAOYSA-N

Application

Versatile ligand for CuI mediated room temperature C-N coupling reactions

Pictograms

Skull and crossbones

Signal Word

Danger

Hazard Statements

Hazard Classifications

Acute Tox. 3 Oral - Eye Irrit. 2 - Skin Irrit. 2 - STOT SE 3

Target Organs

Respiratory system

Storage Class Code

6.1C - Combustible acute toxic Cat.3 / toxic compounds or compounds which causing chronic effects

WGK

WGK 3

Flash Point(F)

Not applicable

Flash Point(C)

Not applicable


Choose from one of the most recent versions:

Certificates of Analysis (COA)

Lot/Batch Number

Don't see the Right Version?

If you require a particular version, you can look up a specific certificate by the Lot or Batch number.

Already Own This Product?

Find documentation for the products that you have recently purchased in the Document Library.

Visit the Document Library

Deping Wang et al.
Chemical communications (Cambridge, England), (14), 1891-1893 (2009-03-26)
2-Pyridinyl beta-ketones were identified as new efficient ligands for CuI-catalysed N-arylation of aliphatic amines at room temperature with great selectivity and substrate scope tolerance.

Our team of scientists has experience in all areas of research including Life Science, Material Science, Chemical Synthesis, Chromatography, Analytical and many others.

Contact Technical Service