Skip to Content
Merck
All Photos(1)

Key Documents

775231

Sigma-Aldrich

Methyl 6-bromo-1H-indazole-5-carboxylate

95%

Synonym(s):

6-Bromoindazole-5-carboxylic acid methyl ester

Sign Into View Organizational & Contract Pricing


About This Item

Empirical Formula (Hill Notation):
C9H7BrN2O2
CAS Number:
Molecular Weight:
255.07
MDL number:
UNSPSC Code:
12352100
PubChem Substance ID:

Assay

95%

form

solid

mp

170-175 °C

functional group

bromo
ester

SMILES string

COC(=O)c1cc2cn[nH]c2cc1Br

InChI

1S/C9H7BrN2O2/c1-14-9(13)6-2-5-4-11-12-8(5)3-7(6)10/h2-4H,1H3,(H,11,12)

InChI key

YKCNVRWLHPWOTP-UHFFFAOYSA-N

Storage Class Code

11 - Combustible Solids

WGK

WGK 3

Flash Point(F)

Not applicable

Flash Point(C)

Not applicable


Choose from one of the most recent versions:

Certificates of Analysis (COA)

Lot/Batch Number

Don't see the Right Version?

If you require a particular version, you can look up a specific certificate by the Lot or Batch number.

Already Own This Product?

Find documentation for the products that you have recently purchased in the Document Library.

Visit the Document Library

Questions

Reviews

No rating value

Active Filters

Our team of scientists has experience in all areas of research including Life Science, Material Science, Chemical Synthesis, Chromatography, Analytical and many others.

Contact Technical Service