Skip to Content
Merck
All Photos(2)

Documents

772208

Sigma-Aldrich

Allyl α-D-mannopyranoside

95%

Synonym(s):

2-Propenyl α-D-mannopyranoside

Sign Into View Organizational & Contract Pricing


About This Item

Empirical Formula (Hill Notation):
C9H16O6
CAS Number:
Molecular Weight:
220.22
MDL number:
UNSPSC Code:
12352201
PubChem Substance ID:
NACRES:
NA.23

Assay

95%

form

solid

mp

94-100 °C

storage temp.

2-8°C

SMILES string

OC[C@H]1O[C@H](OCC=C)[C@@H](O)[C@@H](O)[C@@H]1O

InChI

1S/C9H16O6/c1-2-3-14-9-8(13)7(12)6(11)5(4-10)15-9/h2,5-13H,1,3-4H2/t5-,6-,7+,8+,9+/m1/s1

InChI key

XJNKZTHFPGIJNS-DFTQBPQZSA-N

Storage Class Code

11 - Combustible Solids

WGK

WGK 3

Flash Point(F)

Not applicable

Flash Point(C)

Not applicable


Certificates of Analysis (COA)

Search for Certificates of Analysis (COA) by entering the products Lot/Batch Number. Lot and Batch Numbers can be found on a product’s label following the words ‘Lot’ or ‘Batch’.

Already Own This Product?

Find documentation for the products that you have recently purchased in the Document Library.

Visit the Document Library

Our team of scientists has experience in all areas of research including Life Science, Material Science, Chemical Synthesis, Chromatography, Analytical and many others.

Contact Technical Service