Skip to Content
Merck
All Photos(1)

Documents

766437

Sigma-Aldrich

2-[Di(2-methoxyphenyl)phosphino]benzenesulfonic acid

97%

Sign Into View Organizational & Contract Pricing


About This Item

Empirical Formula (Hill Notation):
C20H19O5PS
CAS Number:
Molecular Weight:
402.40
MDL number:
UNSPSC Code:
12352112
PubChem Substance ID:
NACRES:
NA.22

Assay

97%

form

solid

reaction suitability

reaction type: Buchwald-Hartwig Cross Coupling Reaction
reaction type: Heck Reaction
reaction type: Hiyama Coupling
reaction type: Negishi Coupling
reaction type: Sonogashira Coupling
reaction type: Stille Coupling
reaction type: Suzuki-Miyaura Coupling
reagent type: ligand

mp

120-133 °C

functional group

phosphine

SMILES string

COc1ccccc1P(c2ccccc2OC)c3ccccc3S(O)(=O)=O

InChI

1S/C20H19O5PS/c1-24-15-9-3-5-11-17(15)26(18-12-6-4-10-16(18)25-2)19-13-7-8-14-20(19)27(21,22)23/h3-14H,1-2H3,(H,21,22,23)

InChI key

QFWGCBLAZRYGIG-UHFFFAOYSA-N

Application

Phosphinosulfonate ligand used with Pd and Ni to make polymerization catalysts.

Storage Class Code

11 - Combustible Solids

WGK

WGK 3

Flash Point(F)

Not applicable

Flash Point(C)

Not applicable


Certificates of Analysis (COA)

Search for Certificates of Analysis (COA) by entering the products Lot/Batch Number. Lot and Batch Numbers can be found on a product’s label following the words ‘Lot’ or ‘Batch’.

Already Own This Product?

Find documentation for the products that you have recently purchased in the Document Library.

Visit the Document Library

Damien Guironnet et al.
Journal of the American Chemical Society, 132(12), 4418-4426 (2010-03-09)
Complexes [{(PwedgeO)PdMe}(n)] (1(n); PwedgeO = kappa(2)-P,O-Ar(2)PC(6)H(4)SO(2)O with Ar = 2-MeOC(6)H(4)) are a single-component precursor of the (PwedgeO)PdMe fragment devoid of additional coordinating ligands, which also promotes the catalytic oligomerization of acrylates. Exposure of 1(n) to methyl acrylate afforded the two

Our team of scientists has experience in all areas of research including Life Science, Material Science, Chemical Synthesis, Chromatography, Analytical and many others.

Contact Technical Service